Search
Search

Inha| Gene Symbol: Inha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510601.2 | XP_003510649.1 | 1 - 2 | 326 | 114 | |
| XM_007651222.1 | XP_007649412.1 | 2 - 3 | 429 | 114 | ||
| XM_007651223.1 | XP_007649413.1 | 2 - 3 | 363 | 114 | ||
Set Membership: |
|