Search
Search

Ca12| Gene Symbol: Ca12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499339.2 | XP_003499387.1 | 10 - 11 | 992 | 74 | |
| XM_007640106.1 | XP_007638296.1 | 10 - 11 | 995 | 74 | ||
| XM_007640108.1 | XP_007638298.1 | 9 - 10 | 962 | 74 | ||
Set Membership: |
|