Search
Search

LOC100766798| Gene Symbol: LOC100766798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502500.2 | XP_003502548.2 | 1 - 2 | 67 | 101 | |
| XM_007643114.1 | XP_007641304.1 | 1 - 2 | 180 | 101 | ||
| XM_007643116.1 | XP_007641306.1 | 1 - 2 | 176 | 101 | ||
Set Membership: |
|