Search
Search

Endov| Gene Symbol: Endov | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496871.2 | XP_003496919.1 | 6 - 7 | 604 | 59 | |
| XM_007653939.1 | XP_007652129.1 | 6 - 7 | 568 | 59 | ||
| XM_007653942.1 | XP_007652132.1 | 5 - 6 | 468 | 59 | ||
Set Membership: |
|