Search
Search

Wiz| Gene Symbol: Wiz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504701.2 | XP_003504749.1 | 3 - 4 | 851 | 102 | |
| XM_007645218.1 | XP_007643408.1 | 5 - 6 | 3041 | 102 | ||
| XM_007645222.1 | XP_007643412.1 | 5 - 6 | 3041 | 102 | ||
| GenBank mRNA | JI876625.1 | - | 1 - 2 | 169 | 102 | |
| JP043787.1 | - | 1 - 2 | 169 | 102 | ||
Set Membership: |
|