Search
Search

Fbxl8| Gene Symbol: Fbxl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508536.2 | XP_003508584.1 | 2 - 3 | 266 | 59 | |
| XM_007649154.1 | XP_007647344.1 | 3 - 4 | 400 | 59 | ||
| XM_007649155.1 | XP_007647345.1 | 1 - 2 | 205 | 59 | ||
| XM_007649156.1 | XP_007647346.1 | 2 - 3 | 234 | 59 | ||
| GenBank mRNA | JI895358.1 | - | 2 - 3 | 396 | 59 | |
| JP040135.1 | - | 2 - 3 | 397 | 59 | ||
Set Membership: |
|