Search
Search

Chit1| Gene Symbol: Chit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498841.1 | XP_003498889.1 | 2 - 3 | 57 | 71 | |
| XM_007634173.1 | XP_007632363.1 | 2 - 3 | 57 | 71 | ||
| XM_007634177.1 | XP_007632367.1 | 2 - 3 | 57 | 71 | ||
Set Membership: |
|