Search
Search

Antxr2| Gene Symbol: Antxr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509162.2 | XP_003509210.1 | 15 - 16 | 2296 | 62 | |
| XM_007649768.1 | XP_007647958.1 | 15 - 16 | 2298 | 62 | ||
| GenBank mRNA | FN422397.1 | - | 15 - 16 | 1326 | 62 | |
| JI879406.1 | - | 14 - 15 | 1198 | 62 | ||
| JP040845.1 | - | 14 - 15 | 1198 | 62 | ||
Set Membership: |
|