Search
Search

Reep4| Gene Symbol: Reep4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496160.2 | XP_003496208.1 | 7 - 8 | 1172 | 64 | |
| GenBank mRNA | JI873794.1 | - | 7 - 8 | 1130 | 64 | |
| JP058517.1 | - | 7 - 8 | 1129 | 64 | ||
Set Membership: |
|