Search
Search

Ccdc57| Gene Symbol: Ccdc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654255.1 | XP_007652445.1 | 2 - 3 | 574 | 78 | |
| XM_007654258.1 | XP_007652448.1 | 2 - 3 | 575 | 78 | ||
| XM_007654261.1 | XP_007652451.1 | 2 - 3 | 575 | 78 | ||
| XM_007654267.1 | XP_007652457.1 | 2 - 3 | 574 | 78 | ||