Search
Search

Ccdc57| Gene Symbol: Ccdc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654255.1 | XP_007652445.1 | 5 - 6 | 949 | 101 | |
| XM_007654258.1 | XP_007652448.1 | 5 - 6 | 950 | 101 | ||
| XM_007654261.1 | XP_007652451.1 | 5 - 6 | 950 | 101 | ||
| XM_007654267.1 | XP_007652457.1 | 5 - 6 | 949 | 101 | ||
Set Membership: |
|