Search
Search

Nme1| Gene Symbol: Nme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501376.2 | XP_003501424.1 | 2 - 3 | 232 | 82 | |
| XM_007642024.1 | XP_007640214.1 | 2 - 3 | 150 | 82 | ||
| GenBank mRNA | JI885523.1 | - | 2 - 3 | 188 | 82 | |
| JP038532.1 | - | 3 - 4 | 329 | 82 | ||
Set Membership: |
|