Search
Search

Golga5| Gene Symbol: Golga5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509131.2 | XP_003509179.1 | 11 - 12 | 2157 | 87 | |
| XM_007649732.1 | XP_007647922.1 | 9 - 10 | 1825 | 87 | ||
| GenBank mRNA | JI875028.1 | - | 4 - 5 | 487 | 87 | |
| JP038980.1 | - | 4 - 5 | 492 | 87 | ||
Set Membership: |
|