Search
Search

Med23| Gene Symbol: Med23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513090.2 | XP_003513138.1 | 18 - 19 | 2222 | 62 | |
| XM_003513091.2 | XP_003513139.1 | 17 - 18 | 2204 | 62 | ||
| GenBank mRNA | JI893653.1 | - | 14 - 15 | 1902 | 61 | |
| JP063583.1 | - | 14 - 15 | 1902 | 62 | ||
Set Membership: |
|