Search
Search

Sec23b| Gene Symbol: Sec23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650348.1 | XP_007648538.1 | 10 - 11 | 1336 | 61 | |
| GenBank mRNA | JI869697.1 | - | 10 - 11 | 1330 | 61 | |
| JP042194.1 | - | 10 - 11 | 1330 | 61 | ||
Set Membership: |
|