Search
Search

Map1lc3b| Gene Symbol: Map1lc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495104.2 | XP_003495152.1 | 2 - 3 | 173 | 86 | |
| GenBank mRNA | JI896830.1 | - | 2 - 3 | 181 | 86 | |
| JP037804.1 | - | 2 - 3 | 181 | 86 | ||
Set Membership: |
|