Search
Search

Mrpl37| Gene Symbol: Mrpl37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495759.2 | XP_003495807.1 | 3 - 4 | 705 | 66 | |
| GenBank mRNA | JI888810.1 | - | 2 - 3 | 296 | 66 | |
| JP061135.1 | - | 3 - 4 | 547 | 66 | ||
Set Membership: |
|