Search
Search

Ndufs2| Gene Symbol: Ndufs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647401.1 | XP_007645591.1 | 2 - 3 | 259 | 64 | |
| GenBank mRNA | JI873099.1 | - | 2 - 3 | 411 | 64 | |
| JP045234.1 | - | 2 - 3 | 411 | 64 | ||
Set Membership: |
|