Search
Search

Rps3a| Gene Symbol: Rps3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504173.2 | XP_003504221.1 | 4 - 5 | 637 | 91 | |
| GenBank mRNA | JI885417.1 | - | 4 - 5 | 624 | 91 | |
| JP062748.1 | - | 4 - 5 | 620 | 91 | ||
Set Membership: |
|