Search
Search

Dpy30| Gene Symbol: Dpy30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502489.2 | XP_003502537.1 | 4 - 5 | 346 | 78 | |
| GenBank mRNA | JI888402.1 | - | 4 - 5 | 336 | 78 | |
| JP059280.1 | - | 4 - 5 | 336 | 78 | ||
Set Membership: |
|