Search
Search

Fuz| Gene Symbol: Fuz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510895.2 | XP_003510943.1 | 2 - 3 | 395 | 57 | |
| XM_007651461.1 | XP_007649651.1 | 2 - 3 | 395 | 57 | ||
| GenBank mRNA | JI881413.1 | - | 2 - 3 | 124 | 57 | |
Set Membership: |
|