Search
Search

LOC100766125| Gene Symbol: LOC100766125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506256.2 | XP_003506304.2 | 12 - 13 | 1563 | 70 | |
| XM_007646754.1 | XP_007644944.1 | 12 - 13 | 1572 | 70 | ||
Set Membership: |
|