Search
Search

Fhl2| Gene Symbol: Fhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640279.1 | XP_007638469.1 | 4 - 5 | 710 | 86 | |
| GenBank mRNA | JI885984.1 | - | 5 - 6 | 789 | 86 | |
| JP060940.1 | - | 5 - 6 | 789 | 86 | ||
Set Membership: |
|