Search
Search

Znf18| Gene Symbol: Znf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497497.2 | XP_003497545.1 | 2 - 3 | 513 | 64 | |
| XM_007613105.1 | XP_007611295.1 | 2 - 3 | 666 | 64 | ||
| XM_007613121.1 | XP_007611311.1 | 2 - 3 | 668 | 64 | ||
Set Membership: |
|