Search
Search

Osbpl7| Gene Symbol: Osbpl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499570.2 | XP_003499618.2 | 13 - 14 | 1475 | 55 | |
| XM_007640327.1 | XP_007638517.1 | 14 - 15 | 1701 | 55 | ||
| XM_007640328.1 | XP_007638518.1 | 13 - 14 | 1786 | 55 | ||
| XM_007640329.1 | XP_007638519.1 | 14 - 15 | 1701 | 55 | ||
Set Membership: |
|