Search
Search

LOC100756942| Gene Symbol: LOC100756942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506295.2 | XP_003506343.2 | 1 - 2 | 689 | 104 | |
| GenBank mRNA | JI891788.1 | - | 1 - 2 | 211 | 104 | |
| JP046642.1 | - | 1 - 2 | 211 | 104 | ||
Set Membership: |
|