Search
Search

Ago3| Gene Symbol: Ago3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500893.2 | XP_003500941.1 | 8 - 9 | 1267 | 61 | |
| XM_007641613.1 | XP_007639803.1 | 4 - 5 | 1879 | 61 | ||
| XM_007641614.1 | XP_007639804.1 | 4 - 5 | 1874 | 61 | ||
Set Membership: |
|