Search
Search

Magix| Gene Symbol: Magix | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505084.2 | XP_003505132.1 | 5 - 6 | 687 | 103 | |
| XM_007645611.1 | XP_007643801.1 | 4 - 5 | 589 | 103 | ||
Set Membership: |
|