Search
Search

Mthfd1l| Gene Symbol: Mthfd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495871.2 | XP_003495919.1 | 17 - 18 | 1971 | 57 | |
| GenBank mRNA | JP044775.1 | - | 10 - 11 | 1018 | 57 | |
Set Membership: |
|