Search
Search

Prpf40a| Gene Symbol: Prpf40a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512962.2 | XP_003513010.1 | 8 - 9 | 728 | 82 | |
| XM_007653340.1 | XP_007651530.1 | 8 - 9 | 728 | 82 | ||
| XM_007653341.1 | XP_007651531.1 | 8 - 9 | 602 | 82 | ||
| XM_007653342.1 | XP_007651532.1 | 8 - 9 | 602 | 82 | ||
| GenBank mRNA | JI862238.1 | - | 7 - 8 | 550 | 82 | |
| JP049342.1 | - | 7 - 8 | 550 | 82 | ||