Search
Search

Znf512| Gene Symbol: Znf512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651390.1 | XP_007649580.1 | 13 - 14 | 1513 | 81 | |
| XM_007651397.1 | XP_007649587.1 | 13 - 14 | 1510 | 81 | ||
| XM_007651402.1 | XP_007649592.1 | 13 - 14 | 1507 | 81 | ||
| XM_007651406.1 | XP_007649596.1 | 13 - 14 | 1504 | 81 | ||
| XM_007651410.1 | XP_007649600.1 | 12 - 13 | 1453 | 81 | ||
| XM_007651414.1 | XP_007649604.1 | 12 - 13 | 1450 | 81 | ||
Set Membership: |
|