Search
Search

Sptbn1| Gene Symbol: Sptbn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503968.2 | XP_003504016.1 | 31 - 32 | 6389 | 64 | |
| GenBank mRNA | JI870266.1 | - | 5 - 6 | 508 | 64 | |
| JP050006.1 | - | 5 - 6 | 508 | 64 | ||
Set Membership: |
|