Search
Search

Phtf2| Gene Symbol: Phtf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504432.2 | XP_003504480.1 | 3 - 4 | 205 | 69 | |
| XM_007644989.1 | XP_007643179.1 | 3 - 4 | 205 | 69 | ||
| GenBank mRNA | JI879443.1 | - | 3 - 4 | 286 | 69 | |
Set Membership: |
|