Search
Search

Sp3| Gene Symbol: Sp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645643.1 | XP_007643833.1 | 3 - 4 | 1761 | 107 | |
| XM_007645651.1 | XP_007643841.1 | 3 - 4 | 1761 | 107 | ||
| GenBank mRNA | JP064027.1 | - | 3 - 4 | 1623 | 107 | |
Set Membership: |
|