Search
Search

Ankdd1b| Gene Symbol: Ankdd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513281.2 | XP_003513329.2 | 13 - 14 | 1553 | 57 | |
| XM_007653581.1 | XP_007651771.1 | 13 - 14 | 1896 | 57 | ||
| XM_007653582.1 | XP_007651772.1 | 13 - 14 | 1839 | 57 | ||
| XM_007653583.1 | XP_007651773.1 | 12 - 13 | 1800 | 57 | ||
| XM_007653584.1 | XP_007651774.1 | 13 - 14 | 1386 | 57 | ||
| XM_007653585.1 | XP_007651775.1 | 12 - 13 | 1743 | 57 | ||
Set Membership: |
|