Search
Search

Atf7ip| Gene Symbol: Atf7ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504422.2 | XP_003504470.2 | 8 - 9 | 2477 | 63 | |
| XM_007644979.1 | XP_007643169.1 | 8 - 9 | 2475 | 63 | ||
| XM_007644980.1 | XP_007643170.1 | 8 - 9 | 2476 | 63 | ||
| XM_007644981.1 | XP_007643171.1 | 8 - 9 | 2476 | 63 | ||
| GenBank mRNA | JI873375.1 | - | 7 - 8 | 2194 | 63 | |
Set Membership: |
|