Search
Search

LOC100756190| Gene Symbol: LOC100756190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511837.2 | XP_003511885.1 | 3 - 4 | 1273 | 84 | |
| XM_007652349.1 | XP_007650539.1 | 4 - 5 | 1503 | 84 | ||
Set Membership: |
|