Search
Search

Caprin1| Gene Symbol: Caprin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007612532.1 | XP_007610722.1 | 16 - 17 | 1854 | 61 | |
| GenBank mRNA | JI867560.1 | - | 16 - 17 | 1853 | 61 | |
Set Membership: |
|