Search
Search

LOC100752268| Gene Symbol: LOC100752268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653648.1 | XP_007651838.1 | 4 - 5 | 348 | 70 | |
| GenBank mRNA | JI891671.1 | - | 348 | 70 | ||
Set Membership: |
|