Search
Search

Strbp| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504474.2 | XP_003504522.1 | 8 - 9 | 1131 | 66 | |
| GenBank mRNA | JI879137.1 | - | 7 - 8 | 1003 | 66 | |
| JP056825.1 | - | 8 - 9 | 1132 | 66 | ||
Set Membership: |
|