Search
Search

Bop1| Gene Symbol: Bop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507666.2 | XP_003507714.1 | 7 - 8 | 1039 | 62 | |
| XM_007648239.1 | XP_007646429.1 | 7 - 8 | 1033 | 62 | ||
| XM_007648240.1 | XP_007646430.1 | 6 - 7 | 827 | 62 | ||
| GenBank mRNA | JI880055.1 | - | 7 - 8 | 965 | 62 | |
| JP045723.1 | - | 7 - 8 | 965 | 62 | ||
Set Membership: |
|