Search
Search

Cpt1a| Gene Symbol: Cpt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510378.2 | XP_003510426.2 | 496 | 63 | ||
| GenBank mRNA | JI890502.1 | - | 496 | 63 | ||
| JP040849.1 | - | 496 | 63 | |||
Set Membership: |
|