Search
Search

LOC100755338| Gene Symbol: LOC100755338 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497387.2 | XP_003497435.1 | 7 - 8 | 1058 | 66 | |
| XM_007611452.1 | XP_007609642.1 | 7 - 8 | 1035 | 66 | ||
| XM_007611459.1 | XP_007609649.1 | 7 - 8 | 1039 | 66 | ||
| XM_007611470.1 | XP_007609660.1 | 7 - 8 | 1035 | 66 | ||
Set Membership: |
|