Search
Search

Dnah17| Gene Symbol: Dnah17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498821.2 | XP_003498869.1 | 71 - 72 | 11671 | 86 | |
| XM_007633821.1 | XP_007632011.1 | 71 - 72 | 11698 | 86 | ||
Set Membership: |
|