Search
Search

Znf157| Gene Symbol: Znf157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512244.2 | XP_003512292.2 | 2 - 3 | 274 | 85 | |
| XM_007652758.1 | XP_007650948.1 | 3 - 4 | 377 | 85 | ||
| XM_007652760.1 | XP_007650950.1 | 3 - 4 | 503 | 85 | ||
| XM_007652761.1 | XP_007650951.1 | 3 - 4 | 400 | 85 | ||