Search
Search

Cep170| Gene Symbol: Cep170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499838.2 | XP_003499886.1 | 13 - 14 | 3946 | 63 | |
| XM_003499839.2 | XP_003499887.1 | 12 - 13 | 3653 | 63 | ||
| XM_003499840.2 | XP_003499888.1 | 12 - 13 | 3545 | 63 | ||
| XM_007640586.1 | XP_007638776.1 | 13 - 14 | 3947 | 63 | ||
| XM_007640587.1 | XP_007638777.1 | 13 - 14 | 3946 | 63 | ||
| XM_007640589.1 | XP_007638779.1 | 13 - 14 | 3839 | 63 | ||
| XM_007640590.1 | XP_007638780.1 | 12 - 13 | 3653 | 63 | ||
| GenBank mRNA | JI884707.1 | - | 4 - 5 | 2328 | 63 | |
| JP045707.1 | - | 5 - 6 | 2671 | 63 | ||
Set Membership: |
|