Search
Search

LOC100766249| Gene Symbol: LOC100766249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510950.2 | XP_003510998.1 | 1 - 2 | 79 | 100 | |
| XM_003510951.1 | XP_003510999.1 | 1 - 2 | 79 | 100 | ||
| XM_007651518.1 | XP_007649708.1 | 1 - 2 | 79 | 100 | ||
Set Membership: |
|