Search
Search

CACNA1F| Gene Symbol: CACNA1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045135.1 | XP_017900624.1 | 29 - 30 | 3612 | 69 | |
| XM_018045136.1 | XP_017900625.1 | 29 - 30 | 3579 | 69 | ||
| XM_018045137.1 | XP_017900626.1 | 28 - 29 | 3417 | 69 | ||
Set Membership: |
|