Search
Search
| Gene Symbol: RNF38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683871.3 | XP_005683928.1 | 9 - 10 | 1547 | 70 | |
| XM_018052172.1 | XP_017907661.1 | 11 - 12 | 1562 | 70 | ||
| XM_018052173.1 | XP_017907662.1 | 11 - 12 | 1475 | 70 | ||
| XM_018052174.1 | XP_017907663.1 | 10 - 11 | 1412 | 70 | ||
| XM_018052175.1 | XP_017907664.1 | 9 - 10 | 1569 | 70 | ||
| XM_018052176.1 | XP_017907665.1 | 10 - 11 | 1848 | 70 | ||
| XM_018052177.1 | XP_017907666.1 | 9 - 10 | 1371 | 70 | ||
| XM_018052178.1 | XP_017907667.1 | 10 - 11 | 1389 | 70 | ||
Set Membership: |
|