Search
Search
| Gene Symbol: MYSM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678346.3 | XP_005678403.2 | 14 - 15 | 1814 | 100 | |
| XM_005678347.3 | XP_005678404.2 | 14 - 15 | 1814 | 100 | ||
| XM_013962797.2 | XP_013818251.2 | 14 - 15 | 1814 | 100 | ||
| XM_013962798.2 | XP_013818252.2 | 13 - 14 | 1790 | 100 | ||
| XM_018044728.1 | XP_017900217.1 | 14 - 15 | 1791 | 100 | ||
| XM_018044734.1 | XP_017900223.1 | 14 - 15 | 1814 | 100 | ||
| GenBank mRNA | JO322198.2 | - | 1 - 2 | 58 | 100 | |
Set Membership: |
|